Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMYD5
Entrez Gene ID
10322
Gene Name
SMYD family member 5
Gene Aliases
NN8-4AG, RAI15, RRG1, ZMYND23
Chromosome Location
Chr.2: 73214238 - 73227227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006062.2 NP_006053.2 6 - 7 668 82
XM_006711918.3 XP_006711981.1 6 - 7 755 82
XM_017003163.1 XP_016858652.1 5 - 6 2388 82
GenBank mRNA AK300779.1 - 6 - 7 642 82
BC073806.1 - 6 - 7 644 82
U50383.1 - 6 - 7 621 82

View Full Product Details