The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012455.1 | NP_001012458.1 | 3 - 4 | 740 | 63 |
XM_005248950.2 | XP_005249007.1 | 3 - 4 | 738 | 63 | |
XM_011514406.2 | XP_011512708.1 | 3 - 4 | 738 | 63 | |
XM_011514408.2 | XP_011512710.1 | 4 - 5 | 925 | 63 | |
XM_011514410.2 | XP_011512712.1 | 3 - 4 | 738 | 63 | |
XM_017010479.1 | XP_016865968.1 | 3 - 4 | 3954 | 63 | |
GenBank mRNA | AL833347.1 | - | 3 - 4 | 705 | 63 |
BC101557.1 | - | 2 - 3 | 642 | 63 | |
BC101559.1 | - | 2 - 3 | 642 | 63 | |
BX283350.1 | - | 2 - 3 | 256 | 63 |