Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206811.3 NP_996534.2 6 - 7 871 79
GenBank mRNA AY566854.1 - 6 - 7 636 79
U18840.1 - 6 - 7 642 79
U64566.1 - 5 - 6 555 79

View Full Product Details