The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324459.1 | NP_001311388.1 | 1 - 2 | 76 | 107 |
GenBank mRNA | BI828249.1 | - | 1 - 2 | 77 | 108 |