The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024881.4 | NP_079157.3 | 4 - 5 | 768 | 98 |
GenBank mRNA | AK027699.1 | - | 4 - 5 | 562 | 98 |
AK027850.1 | - | 2 - 3 | 179 | 98 | |
AK075355.1 | - | 2 - 3 | 207 | 98 | |
BC062562.1 | - | 4 - 5 | 768 | 98 |