The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242614.1 | NP_001229543.1 | 7 - 8 | 740 | 69 |
NM_031462.3 | NP_113650.2 | 7 - 8 | 728 | 69 | |
NM_134445.3 | NP_604394.1 | 4 - 5 | 512 | 69 | |
NM_134446.3 | NP_604395.1 | 5 - 6 | 581 | 69 | |
XM_017029890.1 | XP_016885379.1 | 7 - 8 | 592 | 69 | |
XM_017029891.1 | XP_016885380.1 | 5 - 6 | 579 | 69 | |
GenBank mRNA | AK023776.1 | - | 6 - 7 | 414 | 69 |
AK024278.1 | - | 3 - 4 | 235 | 69 | |
AK125020.1 | - | 7 - 8 | 551 | 69 | |
AK290200.1 | - | 7 - 8 | 69 | ||
AL136580.1 | - | 7 - 8 | 617 | 69 | |
AY078165.1 | - | 7 - 8 | 536 | 69 | |
AY078166.1 | - | 5 - 6 | 389 | 69 | |
AY078167.1 | - | 4 - 5 | 320 | 69 | |
AY358837.1 | - | 7 - 8 | 601 | 69 | |
BC007569.1 | - | 6 - 7 | 388 | 69 | |
BC013029.2 | - | 6 - 7 | 388 | 69 | |
BC025729.1 | - | 8 - 9 | 1841 | 69 | |
BC030536.1 | - | 7 - 8 | 588 | 69 | |
KU178797.1 | - | 7 - 8 | 500 | 69 |