The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000095.2 | NP_000086.2 | 16 - 17 | 1951 | 95 |
GenBank mRNA | AB086984.1 | - | 16 - 17 | 1915 | 95 |
AK223216.1 | - | 16 - 17 | 1951 | 95 | |
AK290595.1 | - | 16 - 17 | 1951 | 95 | |
AK296586.1 | - | 15 - 16 | 1792 | 95 | |
AK297798.1 | - | 15 - 16 | 1853 | 95 | |
AK297814.1 | - | 15 - 16 | 1912 | 95 | |
BC033676.1 | - | 15 - 16 | 2175 | 95 | |
BC110847.1 | - | 16 - 17 | 1937 | 95 | |
BC125092.1 | - | 16 - 17 | 1944 | 95 | |
L32137.1 | - | 16 - 17 | 1940 | 95 |