Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMP
Entrez Gene ID
1311
Gene Name
cartilage oligomeric matrix protein
Gene Aliases
EDM1, EPD1, MED, PSACH, THBS5, TSP5
Chromosome Location
Chr.19: 18782773 - 18791305 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000095.2 NP_000086.2 2 - 3 200 109
GenBank mRNA AB086984.1 - 2 - 3 109
AK074508.1 - 2 - 3 200 109
AK223216.1 - 2 - 3 200 109
AK290595.1 - 2 - 3 200 109
AK296586.1 - 2 - 3 200 109
AK297814.1 - 2 - 3 200 109
AU120397.1 - 2 - 3 200 109
BC033676.1 - 1 - 2 424 109
BC110847.1 - 2 - 3 186 109
BC125092.1 - 2 - 3 193 109
L32137.1 - 2 - 3 109

View Full Product Details