Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SP2
Entrez Gene ID
6668
Gene Name
Sp2 transcription factor
Gene Aliases
-
Chromosome Location
Chr.17: 47896032 - 47932271 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006722025.2 XP_006722088.1 1 - 2 70 86
XM_006722026.2 XP_006722089.1 1 - 2 116 86
XM_011525142.1 XP_011523444.1 1 - 2 70 86

View Full Product Details