The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164712.1 | NP_001158184.1 | 9 - 10 | 1365 | 68 |
GenBank mRNA | BC007546.1 | - | 9 - 10 | 1129 | 68 |