The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166461.1 | NP_001159933.1 | 5 - 6 | 633 | 58 |
GenBank mRNA | AY634686.1 | - | 5 - 6 | 503 | 58 |