The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201338.1 | NP_001188267.1 | 19 - 20 | 2732 | 61 |
NM_001201339.1 | NP_001188268.1 | 19 - 20 | 2729 | 61 | |
NM_001201340.1 | NP_001188269.1 | 18 - 19 | 2525 | 61 | |
NM_001320571.1 | NP_001307500.1 | 17 - 18 | 2457 | 61 | |
NM_001320572.1 | NP_001307501.1 | 19 - 20 | 2723 | 61 | |
NM_002967.3 | NP_002958.2 | 19 - 20 | 2726 | 61 | |
XM_017027114.1 | XP_016882603.1 | 17 - 18 | 2414 | 61 | |
XM_017027115.1 | XP_016882604.1 | 17 - 18 | 2408 | 61 | |
XM_017027116.1 | XP_016882605.1 | 18 - 19 | 2122 | 61 | |
XM_017027117.1 | XP_016882606.1 | 18 - 19 | 2119 | 61 | |
XM_017027118.1 | XP_016882607.1 | 18 - 19 | 2113 | 61 | |
GenBank mRNA | AK095736.1 | - | 15 - 16 | 1908 | 61 |
AK290444.1 | - | 19 - 20 | 2625 | 61 | |
AK293152.1 | - | 16 - 17 | 2230 | 61 | |
AK294671.1 | - | 17 - 18 | 2182 | 61 | |
AK295298.1 | - | 16 - 17 | 2315 | 61 | |
AK298707.1 | - | 18 - 19 | 2421 | 61 | |
AK299525.1 | - | 19 - 20 | 2490 | 61 | |
AK304474.1 | - | 15 - 16 | 1880 | 61 | |
BC126219.1 | - | 19 - 20 | 2603 | 61 | |
BC143937.1 | - | 19 - 20 | 2600 | 61 | |
BC143938.1 | - | 19 - 20 | 2594 | 61 | |
BC143939.1 | - | 19 - 20 | 2600 | 61 | |
CN271637.1 | - | 1 - 2 | 99 | 61 | |
L43631.1 | - | 19 - 20 | 2403 | 61 | |
U72355.1 | - | 19 - 20 | 2608 | 61 |