The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000535.6 | NP_000526.2 | 9 - 10 | 1072 | 117 |
NM_001322003.1 | NP_001308932.1 | 9 - 10 | 1067 | 117 | |
NM_001322004.1 | NP_001308933.1 | 8 - 9 | 932 | 117 | |
NM_001322005.1 | NP_001308934.1 | 9 - 10 | 1184 | 117 | |
NM_001322007.1 | NP_001308936.1 | 8 - 9 | 969 | 117 | |
NM_001322008.1 | NP_001308937.1 | 7 - 8 | 829 | 117 | |
NM_001322009.1 | NP_001308938.1 | 9 - 10 | 1179 | 117 | |
NM_001322011.1 | NP_001308940.1 | 9 - 10 | 1023 | 117 | |
NM_001322012.1 | NP_001308941.1 | 9 - 10 | 1018 | 117 | |
NM_001322013.1 | NP_001308942.1 | 8 - 9 | 899 | 117 | |
NM_001322014.1 | NP_001308943.1 | 9 - 10 | 1072 | 117 | |
NM_001322015.1 | NP_001308944.1 | 9 - 10 | 1242 | 117 | |
NR_136154.1 | 9 - 10 | 1072 | 117 | ||
XM_006715744.3 | XP_006715807.1 | 2 - 3 | 148 | 117 | |
XM_017012342.1 | XP_016867831.1 | 8 - 9 | 897 | 117 | |
GenBank mRNA | AB103082.1 | - | 9 - 10 | 985 | 117 |
AB103086.1 | - | 6 - 7 | 667 | 117 | |
AK294661.1 | - | 8 - 9 | 922 | 117 | |
AK312390.1 | - | 9 - 10 | 1021 | 117 | |
BC008400.1 | - | 9 - 10 | 1030 | 117 | |
BC031832.1 | - | 9 - 10 | 1036 | 117 | |
BC093921.1 | - | 9 - 10 | 1007 | 117 | |
U14658.1 | - | 9 - 10 | 987 | 117 |