Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC2B
Entrez Gene ID
9976
Gene Name
C-type lectin domain family 2 member B
Gene Aliases
AICL, CLECSF2, HP10085, IFNRG1
Chromosome Location
Chr.12: 9852369 - 9869859 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005127.2 NP_005118.2 2 - 3 734 140
GenBank mRNA AB015628.1 - 2 - 3 227 140
AK307334.1 - 2 - 3 261 140
BC005254.1 - 2 - 3 228 140
X96719.1 - 2 - 3 209 140

View Full Product Details