The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040147.2 | NP_001035237.1 | 3 - 4 | 312 | 103 |
NM_001261830.1 | NP_001248759.1 | 3 - 4 | 287 | 103 | |
NM_003784.3 | NP_003775.1 | 3 - 4 | 580 | 103 | |
GenBank mRNA | AF027866.1 | - | 3 - 4 | 584 | 103 |
AK290673.1 | - | 3 - 4 | 284 | 103 | |
AK290876.1 | - | 3 - 4 | 531 | 103 | |
BC069417.1 | - | 3 - 4 | 532 | 103 | |
BC069442.1 | - | 3 - 4 | 532 | 103 | |
BC069547.1 | - | 3 - 4 | 532 | 103 | |
BC106743.2 | - | 3 - 4 | 440 | 103 | |
BC106744.2 | - | 3 - 4 | 440 | 103 | |
CD702645.1 | - | 3 - 4 | 342 | 103 | |
D88575.1 | - | 3 - 4 | 293 | 103 | |
DA577978.1 | - | 3 - 4 | 288 | 103 |