Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTSA
Entrez Gene ID
5476
Gene Name
cathepsin A
Gene Aliases
GLB2, GSL, NGBE, PPCA, PPGB
Chromosome Location
Chr.20: 45890144 - 45898820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000308.3 NP_000299.2 6 - 7 1835 73
NM_001127695.2 NP_001121167.1 6 - 7 1881 73
NM_001167594.2 NP_001161066.1 5 - 6 1784 73
GenBank mRNA AB209705.2 - 6 - 7 651 73
AK097786.1 - 5 - 6 548 73
AK304536.1 - 5 - 6 612 73
AK307701.1 - 6 - 7 628 73
AK312898.1 - 6 - 7 644 73
BC000597.2 - 6 - 7 637 73
BC093009.1 - 6 - 7 644 73
M22960.1 - 5 - 6 605 73

View Full Product Details