Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTBDN
Entrez Gene ID
83546
Gene Name
retbindin
Gene Aliases
-
Chromosome Location
Chr.19: 12825477 - 12835428 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080997.2 NP_001074466.1 5 - 6 611 95
NM_001270441.1 NP_001257370.1 5 - 6 564 95
NM_001270442.1 NP_001257371.1 5 - 6 552 95
NM_001270443.1 NP_001257372.1 5 - 6 540 95
NM_001270444.1 NP_001257373.1 5 - 6 528 95
NM_001270445.1 NP_001257374.1 5 - 6 522 95
NM_031429.2 NP_113617.1 6 - 7 884 95
XM_005260089.1 XP_005260146.1 6 - 7 959 95
XM_005260090.3 XP_005260147.1 5 - 6 822 95
XM_011528343.1 XP_011526645.1 5 - 6 625 95
XM_011528344.2 XP_011526646.1 5 - 6 675 95
XM_011528345.1 XP_011526647.1 5 - 6 620 95
XM_017027343.1 XP_016882832.1 6 - 7 914 95
XM_017027344.1 XP_016882833.1 6 - 7 866 95
GenBank mRNA AB593122.1 - 5 - 6 564 95
AY028917.1 - 5 - 6 588 95
BC005063.1 - 6 - 7 872 95
BG471112.1 - 5 - 6 525 95
BM688006.1 - 5 - 6 516 95
BQ639040.1 - 6 - 7 486 95
BU689643.1 - 1 - 2 95

View Full Product Details