The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018465.3 | NP_060935.2 | 5 - 6 | 610 | 83 |
XM_005251510.4 | XP_005251567.1 | 5 - 6 | 725 | 83 | |
XM_005251512.4 | XP_005251569.1 | 3 - 4 | 398 | 83 | |
XM_011517960.2 | XP_011516262.1 | 5 - 6 | 758 | 83 | |
GenBank mRNA | AF220050.1 | - | 5 - 6 | 534 | 83 |
AF225420.1 | - | 5 - 6 | 551 | 83 | |
AK027594.1 | - | 5 - 6 | 549 | 83 | |
AK312736.1 | - | 5 - 6 | 539 | 83 | |
BC008212.1 | - | 5 - 6 | 520 | 83 |