The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282962.1 | NP_001269891.1 | 1 - 2 | 185 | 71 |
NM_001282963.1 | NP_001269892.1 | 1 - 2 | 185 | 71 | |
NM_018410.4 | NP_060880.3 | 1 - 2 | 185 | 71 | |
XM_011511437.1 | XP_011509739.1 | 1 - 2 | 154 | 71 | |
GenBank mRNA | AB101211.1 | - | 1 - 2 | 186 | 71 |
AB162218.1 | - | 1 - 2 | 119 | 71 | |
AK074809.1 | - | 1 - 2 | 151 | 71 | |
AK301643.1 | - | 1 - 2 | 154 | 71 | |
AK303109.1 | - | 1 - 2 | 126 | 71 | |
BC001940.1 | - | 1 - 2 | 119 | 71 | |
BM148072.1 | - | 1 - 2 | 186 | 71 | |
BM838206.1 | - | 1 - 2 | 149 | 71 | |
BQ225710.1 | - | 1 - 2 | 141 | 71 | |
DB022872.1 | - | 1 - 2 | 183 | 71 |