Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18BP1
Entrez Gene ID
55320
Gene Name
MIS18 binding protein 1
Gene Aliases
C14orf106, HSA242977, KNL2, M18BP1
Chromosome Location
Chr.14: 45203190 - 45253402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018353.4 NP_060823.3 6 - 7 1806 154
XM_005267833.4 XP_005267890.1 7 - 8 1853 154
XM_011536923.1 XP_011535225.1 3 - 4 291 154
XM_017021425.1 XP_016876914.1 8 - 9 1834 154
GenBank mRNA AB067490.1 - 6 - 7 1807 154
AK002048.1 - 6 - 7 1601 154
BC051885.1 - 6 - 7 1583 154
BC065544.1 - 6 - 7 1597 154

View Full Product Details