The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032840.2 | NP_116229.1 | 2 - 3 | 256 | 88 |
GenBank mRNA | AK027706.1 | - | 2 - 3 | 190 | 88 |
AK074694.1 | - | 2 - 3 | 222 | 88 | |
AK294092.1 | - | 2 - 3 | 240 | 88 | |
BC136312.1 | - | 2 - 3 | 189 | 88 | |
DA389433.1 | - | 2 - 3 | 256 | 88 |