Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
Chromosome Location
Chr.7: 129434433 - 129488399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134336.1 NP_001127808.1 15 - 16 1688 73
NM_020704.2 NP_065755.1 15 - 16 1688 73
XM_006716069.3 XP_006716132.1 15 - 16 1688 73
XM_011516432.2 XP_011514734.1 12 - 13 1336 73
XM_017012470.1 XP_016867959.1 15 - 16 1688 73
XM_017012471.1 XP_016867960.1 9 - 10 1011 73
XM_017012472.1 XP_016867961.1 13 - 14 3153 73
GenBank mRNA AB032996.1 - 15 - 16 1662 73
AK290165.1 - 15 - 16 1688 73
AK302339.1 - 7 - 8 893 73
BC019064.2 - 15 - 16 1675 73

View Full Product Details