Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRIP2
Entrez Gene ID
57464
Gene Name
striatin interacting protein 2
Gene Aliases
FAM40B, FAR11B
Chromosome Location
Chr.7: 129434433 - 129488399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134336.1 NP_001127808.1 5 - 6 574 75
NM_020704.2 NP_065755.1 5 - 6 574 75
XM_006716069.3 XP_006716132.1 5 - 6 574 75
XM_011516432.2 XP_011514734.1 2 - 3 222 75
XM_011516435.1 XP_011514737.1 5 - 6 574 75
XM_017012470.1 XP_016867959.1 5 - 6 574 75
XM_017012472.1 XP_016867961.1 5 - 6 574 75
GenBank mRNA AB032996.1 - 5 - 6 548 75
AK290165.1 - 5 - 6 574 75
BC019064.2 - 5 - 6 561 75
DA409007.1 - 5 - 6 574 75

View Full Product Details