The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002264.2 | NP_001002264.1 | 12 - 13 | 1079 | 130 |
XM_005266596.1 | XP_005266653.1 | 11 - 12 | 1046 | 130 | |
XM_011535312.1 | XP_011533614.1 | 12 - 13 | 2289 | 130 | |
XM_011535313.1 | XP_011533615.1 | 11 - 12 | 984 | 130 | |
XM_011535314.1 | XP_011533616.1 | 7 - 8 | 481 | 130 | |
XM_017020853.1 | XP_016876342.1 | 12 - 13 | 869 | 130 | |
XM_017020854.1 | XP_016876343.1 | 13 - 14 | 1455 | 130 | |
XM_017020855.1 | XP_016876344.1 | 11 - 12 | 1668 | 130 | |
GenBank mRNA | BC023660.2 | - | 12 - 13 | 1038 | 130 |