The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002264.2 | NP_001002264.1 | 6 - 7 | 665 | 132 |
NM_033255.3 | NP_150280.1 | 6 - 7 | 665 | 132 | |
XM_005266596.1 | XP_005266653.1 | 6 - 7 | 665 | 132 | |
XM_005266597.1 | XP_005266654.1 | 6 - 7 | 665 | 132 | |
XM_006719896.3 | XP_006719959.1 | 6 - 7 | 665 | 132 | |
XM_011535312.1 | XP_011533614.1 | 6 - 7 | 1875 | 132 | |
XM_011535313.1 | XP_011533615.1 | 5 - 6 | 570 | 132 | |
XM_017020853.1 | XP_016876342.1 | 6 - 7 | 455 | 132 | |
XM_017020854.1 | XP_016876343.1 | 7 - 8 | 1041 | 132 | |
XM_017020855.1 | XP_016876344.1 | 5 - 6 | 1254 | 132 | |
GenBank mRNA | AF396928.1 | - | 6 - 7 | 624 | 132 |
AK292461.1 | - | 6 - 7 | 635 | 132 | |
AK313877.1 | - | 6 - 7 | 629 | 132 | |
AL831953.1 | - | 5 - 6 | 2057 | 132 | |
BC023660.2 | - | 6 - 7 | 624 | 132 |