Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
Chromosome Location
Chr.13: 42886388 - 42992271 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033255.3 NP_150280.1 10 - 11 1015 98
XM_005266597.1 XP_005266654.1 11 - 12 1048 98
GenBank mRNA AF396928.1 - 10 - 11 974 98
AK292461.1 - 10 - 11 985 98
AL831953.1 - 10 - 11 2440 98

View Full Product Details