Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLCN2
Entrez Gene ID
1181
Gene Name
chloride voltage-gated channel 2
Gene Aliases
CIC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8, LKPAT, clC-2
Chromosome Location
Chr.3: 184346185 - 184361651 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171087.2 NP_001164558.1 19 - 20 2343 83
NM_001171088.2 NP_001164559.1 18 - 19 2262 83
NM_001171089.2 NP_001164560.1 19 - 20 2394 83
NM_004366.5 NP_004357.3 19 - 20 2394 83
XM_006713489.1 XP_006713552.1 19 - 20 2346 83
XM_006713490.2 XP_006713553.1 13 - 14 1604 83
XM_011512401.1 XP_011510703.1 19 - 20 2346 83
GenBank mRNA AF026004.1 - 19 - 20 2222 83
AK298952.1 - 19 - 20 2346 83
AK302759.1 - 18 - 19 2214 83
BC021578.1 - 9 - 10 1089 83
BC072004.1 - 19 - 20 2294 83
BX089717.1 - 2 - 3 204 83
KU177981.1 - 6 - 7 794 83
KU177982.1 - 6 - 7 794 83
KU177983.1 - 6 - 7 788 83
KU177984.1 - 6 - 7 722 83
KU177985.1 - 3 - 4 276 83

View Full Product Details