The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171087.2 | NP_001164558.1 | 23 - 24 | 2623 | 65 |
NM_001171088.2 | NP_001164559.1 | 22 - 23 | 2542 | 65 | |
NM_004366.5 | NP_004357.3 | 23 - 24 | 2674 | 65 | |
XM_006713489.1 | XP_006713552.1 | 22 - 23 | 2521 | 65 | |
XM_006713490.2 | XP_006713553.1 | 17 - 18 | 1884 | 65 | |
GenBank mRNA | AF026004.1 | - | 23 - 24 | 2502 | 65 |
AK302759.1 | - | 22 - 23 | 2494 | 65 | |
BC021578.1 | - | 12 - 13 | 1264 | 65 | |
BC072004.1 | - | 23 - 24 | 2574 | 65 | |
BX089717.1 | - | 6 - 7 | 484 | 65 | |
KU177981.1 | - | 9 - 10 | 969 | 65 | |
KU177982.1 | - | 10 - 11 | 1074 | 65 | |
KU177983.1 | - | 10 - 11 | 1068 | 65 | |
KU177984.1 | - | 10 - 11 | 1002 | 65 | |
KU177985.1 | - | 6 - 7 | 451 | 65 |