The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143773.1 | NP_001137245.1 | 10 - 11 | 1324 | 124 |
NM_001166698.1 | NP_001160170.1 | 10 - 11 | 1238 | 124 | |
NM_198215.3 | NP_937858.2 | 9 - 10 | 1153 | 124 | |
XM_005269618.4 | XP_005269675.2 | 9 - 10 | 1153 | 124 | |
XM_006717702.3 | XP_006717765.1 | 11 - 12 | 1154 | 124 | |
XM_006717703.3 | XP_006717766.1 | 11 - 12 | 1336 | 124 | |
XM_011539490.2 | XP_011537792.1 | 10 - 11 | 1209 | 124 | |
XM_011539491.2 | XP_011537793.1 | 11 - 12 | 1255 | 124 | |
XM_017015885.1 | XP_016871374.1 | 10 - 11 | 1067 | 124 | |
XM_017015886.1 | XP_016871375.1 | 10 - 11 | 1324 | 124 | |
XM_017015887.1 | XP_016871376.1 | 10 - 11 | 1074 | 124 | |
XM_017015888.1 | XP_016871377.1 | 11 - 12 | 1336 | 124 | |
XM_017015889.1 | XP_016871378.1 | 11 - 12 | 1154 | 124 | |
XM_017015890.1 | XP_016871379.1 | 11 - 12 | 1393 | 124 | |
GenBank mRNA | AK289796.1 | - | 9 - 10 | 1079 | 124 |
AK294805.1 | - | 9 - 10 | 1068 | 124 | |
AK299924.1 | - | 10 - 11 | 1324 | 124 | |
AK316542.1 | - | 10 - 11 | 1234 | 124 | |
BC036453.1 | - | 9 - 10 | 1111 | 124 | |
U79304.1 | - | 7 - 8 | 761 | 124 |