The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042369.2 | NP_001035828.1 | 3 - 4 | 1164 | 124 |
NM_001042370.2 | NP_001035829.2 | 3 - 4 | 1164 | 124 | |
NM_001173524.1 | NP_001166995.1 | 3 - 4 | 1170 | 124 | |
NM_001173525.1 | NP_001166996.1 | 3 - 4 | 1164 | 124 | |
NM_004600.5 | NP_004591.2 | 3 - 4 | 1164 | 124 | |
NR_033393.1 | 2 - 3 | 569 | 124 | ||
XM_006711495.3 | XP_006711558.1 | 4 - 5 | 1236 | 124 | |
XM_006711496.3 | XP_006711559.1 | 5 - 6 | 1412 | 124 | |
XM_006711497.3 | XP_006711560.1 | 4 - 5 | 1346 | 124 | |
XM_011509922.2 | XP_011508224.1 | 3 - 4 | 745 | 124 | |
XM_017002180.1 | XP_016857669.1 | 4 - 5 | 1045 | 124 | |
XM_017002181.1 | XP_016857670.1 | 4 - 5 | 1274 | 124 | |
XM_017002182.1 | XP_016857671.1 | 3 - 4 | 979 | 124 | |
GenBank mRNA | AK314594.1 | - | 3 - 4 | 957 | 124 |
AY205314.1 | - | 3 - 4 | 978 | 124 | |
AY205315.1 | - | 3 - 4 | 978 | 124 | |
BC036658.2 | - | 3 - 4 | 1006 | 124 | |
CR936683.1 | - | 2 - 3 | 538 | 124 | |
M25077.1 | - | 3 - 4 | 978 | 124 |