The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167738.1 | NP_001161210.1 | 24 - 25 | 4229 | 57 |
NM_020443.4 | NP_065176.3 | 27 - 28 | 5569 | 57 | |
XM_006711609.2 | XP_006711672.2 | 25 - 26 | 4204 | 57 | |
XM_006711610.2 | XP_006711673.2 | 24 - 25 | 4180 | 57 | |
XM_006711611.1 | XP_006711674.1 | 25 - 26 | 4320 | 57 | |
XM_011510097.1 | XP_011508399.1 | 26 - 27 | 5239 | 57 | |
XM_011510098.1 | XP_011508400.1 | 26 - 27 | 5224 | 57 | |
XM_011510099.1 | XP_011508401.1 | 25 - 26 | 5215 | 57 | |
XM_011510100.1 | XP_011508402.1 | 26 - 27 | 5077 | 57 | |
XM_011510101.1 | XP_011508403.1 | 25 - 26 | 5053 | 57 | |
XM_011510102.1 | XP_011508404.1 | 24 - 25 | 5044 | 57 | |
XM_017002751.1 | XP_016858240.1 | 25 - 26 | 5068 | 57 | |
XM_017002752.1 | XP_016858241.1 | 23 - 24 | 4171 | 57 | |
GenBank mRNA | AB032977.3 | - | 27 - 28 | 4902 | 57 |
AJ488101.1 | - | 27 - 28 | 5100 | 57 | |
AK022622.1 | - | 5 - 6 | 834 | 57 | |
AK024265.1 | - | 12 - 13 | 1734 | 57 | |
AY043013.1 | - | 27 - 28 | 5569 | 57 | |
BC007523.1 | - | 13 - 14 | 1851 | 57 | |
BX648460.1 | - | 24 - 25 | 4196 | 57 |