Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
-
Chromosome Location
Chr.1: 55066359 - 55215374 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015306.2 NP_056121.2 33 - 34 3983 96
XM_005270690.3 XP_005270747.1 20 - 21 2024 96
XM_017000831.1 XP_016856320.1 34 - 35 4048 96
XM_017000832.1 XP_016856321.1 34 - 35 4045 96
XM_017000833.1 XP_016856322.1 34 - 35 3540 96
XM_017000834.1 XP_016856323.1 33 - 34 3480 96
XM_017000835.1 XP_016856324.1 34 - 35 4051 96
XM_017000836.1 XP_016856325.1 34 - 35 4051 96
XM_017000837.1 XP_016856326.1 34 - 35 4051 96
GenBank mRNA AK127075.1 - 16 - 17 1728 96
BU156226.1 - 2 - 3 149 96

View Full Product Details