Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
SLX4 | NM_032444.2 | NP_115820.2 | ||
XM_011522715.2 | XP_011521017.1 | |||
XM_017023775.1 | XP_016879264.1 | |||
AK095411.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833128 | Chr16:3525122 - 3690497 on Build GRCh38 | Loss | LOC101929732 DNASE1 NLRC3 SLX4 TRAP1 CLUAP1 |
nsv571255 | Chr16:3563125 - 3589138 on Build GRCh38 | Gain | NLRC3 SLX4 |
nsv517451 | Chr16:3549655 - 3611814 on Build GRCh38 | Loss | LOC101929732 DNASE1 NLRC3 SLX4 |