The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032532.2 | NP_115921.2 | 19 - 20 | 5455 | 91 |
XM_011536190.2 | XP_011534492.1 | 18 - 19 | 5378 | 91 | |
XM_011536191.2 | XP_011534493.1 | 16 - 17 | 5096 | 91 | |
GenBank mRNA | AB058769.2 | - | 17 - 18 | 4923 | 91 |
AJ586132.1 | - | 9 - 10 | 1356 | 91 | |
AL832410.1 | - | 9 - 10 | 1654 | 91 | |
BC146783.1 | - | 17 - 18 | 4923 | 91 | |
BC150607.1 | - | 18 - 19 | 5130 | 91 | |
DQ009660.1 | - | 19 - 20 | 5437 | 91 |