Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNDC1
Entrez Gene ID
84624
Gene Name
fibronectin type III domain containing 1
Gene Aliases
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1
Chromosome Location
Chr.6: 159169397 - 159272108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032532.2 NP_115921.2 6 - 7 965 71
XM_011536190.2 XP_011534492.1 5 - 6 888 71
XM_011536191.2 XP_011534493.1 3 - 4 606 71
GenBank mRNA AB058769.2 - 5 - 6 640 71
BC146783.1 - 5 - 6 640 71
BC150607.1 - 5 - 6 640 71
DQ009660.1 - 6 - 7 965 71

View Full Product Details