The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014728.3 | NP_055543.2 | 13 - 14 | 1980 | 69 |
XM_005274632.3 | XP_005274689.2 | 13 - 14 | 1658 | 69 | |
XM_011545613.2 | XP_011543915.1 | 13 - 14 | 1967 | 69 | |
XM_017029983.1 | XP_016885472.1 | 13 - 14 | 1870 | 69 | |
XM_017029984.1 | XP_016885473.1 | 12 - 13 | 1569 | 69 | |
XM_017029985.1 | XP_016885474.1 | 13 - 14 | 1656 | 69 | |
XM_017029986.1 | XP_016885475.1 | 13 - 14 | 1869 | 69 | |
GenBank mRNA | AK289694.1 | - | 13 - 14 | 1967 | 69 |
BC113700.1 | - | 13 - 14 | 1565 | 69 | |
BC113702.1 | - | 13 - 14 | 1565 | 69 | |
BC143746.1 | - | 14 - 15 | 1662 | 69 |