Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD4
Entrez Gene ID
9758
Gene Name
FERM and PDZ domain containing 4
Gene Aliases
PDZD10, PDZK10
Chromosome Location
Chr.X: 12137329 - 12724523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014728.3 NP_055543.2 6 - 7 1077 115
XM_005274632.3 XP_005274689.2 6 - 7 755 115
XM_011545613.2 XP_011543915.1 6 - 7 1064 115
XM_017029983.1 XP_016885472.1 6 - 7 967 115
XM_017029984.1 XP_016885473.1 6 - 7 756 115
XM_017029985.1 XP_016885474.1 6 - 7 753 115
XM_017029986.1 XP_016885475.1 6 - 7 966 115
GenBank mRNA AB002314.3 - 6 - 7 1077 115
AK289694.1 - 6 - 7 1064 115
BC113700.1 - 6 - 7 662 115
BC113702.1 - 6 - 7 662 115
BC143746.1 - 7 - 8 759 115

View Full Product Details