Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
Chromosome Location
Chr.3: 42704118 - 42773253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144719.3 NP_653320.3 11 - 12 1592 80
XM_005264898.3 XP_005264955.1 11 - 12 1592 80
XM_011533419.2 XP_011531721.1 7 - 8 1603 80
XM_017005781.1 XP_016861270.1 7 - 8 990 80
GenBank mRNA BC036050.1 - 11 - 12 1594 80

View Full Product Details