Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
Chromosome Location
Chr.1: 15617458 - 15661057 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK057374.1 - 2 - 3 288 73

View Full Product Details