Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
Chromosome Location
Chr.9: 15552874 - 16062431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173550.2 NP_775821.2 4 - 5 662 74
XM_005251397.2 XP_005251454.1 4 - 5 545 74
XM_005251398.3 XP_005251455.1 4 - 5 784 74
XM_005251399.4 XP_005251456.1 5 - 6 844 74
XM_011517788.2 XP_011516090.1 4 - 5 774 74
XM_011517791.2 XP_011516093.1 4 - 5 784 74
XM_011517796.2 XP_011516098.1 2 - 3 191 74
XM_017014431.1 XP_016869920.1 4 - 5 784 74
XM_017014432.1 XP_016869921.1 4 - 5 784 74
XM_017014433.1 XP_016869922.1 4 - 5 784 74
XM_017014434.1 XP_016869923.1 4 - 5 784 74
XM_017014435.1 XP_016869924.1 4 - 5 784 74
XM_017014436.1 XP_016869925.1 4 - 5 784 74
XM_017014437.1 XP_016869926.1 4 - 5 784 74
XM_017014438.1 XP_016869927.1 4 - 5 783 74
XM_017014441.1 XP_016869930.1 4 - 5 783 74
XM_017014442.1 XP_016869931.1 4 - 5 783 74
XM_017014443.1 XP_016869932.1 2 - 3 186 74
GenBank mRNA AK308284.1 - 4 - 5 545 74
AY422473.1 - 4 - 5 662 74
BC144214.1 - 3 - 4 439 74
BC150600.1 - 3 - 4 439 74
BX537933.1 - 4 - 5 523 74

View Full Product Details