The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 10 - 11 | 1600 | 102 |
XM_011542067.2 | XP_011540369.1 | 11 - 12 | 2532 | 102 | |
XM_011542068.2 | XP_011540370.1 | 11 - 12 | 2537 | 102 | |
XM_011542069.2 | XP_011540371.1 | 10 - 11 | 2391 | 102 | |
XM_011542070.2 | XP_011540372.1 | 9 - 10 | 1558 | 102 | |
XM_011542071.2 | XP_011540373.1 | 8 - 9 | 1967 | 102 | |
XM_017002225.1 | XP_016857714.1 | 10 - 11 | 1617 | 102 | |
XM_017002226.1 | XP_016857715.1 | 11 - 12 | 1928 | 102 | |
GenBank mRNA | AL832217.1 | - | 17 - 18 | 2633 | 102 |
AY639646.1 | - | 9 - 10 | 1425 | 102 |