The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 13 - 14 | 1995 | 93 |
XM_011542067.2 | XP_011540369.1 | 14 - 15 | 2927 | 93 | |
XM_011542068.2 | XP_011540370.1 | 14 - 15 | 2932 | 93 | |
XM_011542069.2 | XP_011540371.1 | 13 - 14 | 2786 | 93 | |
XM_011542070.2 | XP_011540372.1 | 12 - 13 | 1953 | 93 | |
XM_011542071.2 | XP_011540373.1 | 11 - 12 | 2362 | 93 | |
XM_017002225.1 | XP_016857714.1 | 13 - 14 | 2012 | 93 | |
XM_017002226.1 | XP_016857715.1 | 14 - 15 | 2323 | 93 | |
GenBank mRNA | AL832217.1 | - | 20 - 21 | 3028 | 93 |
AY639646.1 | - | 12 - 13 | 1820 | 93 |