The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011542069.2 | XP_011540371.1 | 3 - 4 | 1160 | 94 |
GenBank mRNA | AK302075.1 | - | 5 - 6 | 553 | 94 |
AL832217.1 | - | 9 - 10 | 1264 | 94 |