The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024646.2 | NP_078922.1 | 13 - 14 | 2240 | 69 |
XM_006710898.3 | XP_006710961.1 | 13 - 14 | 2127 | 69 | |
GenBank mRNA | AB051517.1 | - | 13 - 14 | 2190 | 69 |
AK056645.1 | - | 11 - 12 | 1184 | 69 | |
AK095643.1 | - | 12 - 13 | 1880 | 69 | |
AK296460.1 | - | 11 - 12 | 2035 | 69 | |
BC029832.1 | - | 11 - 12 | 1254 | 69 | |
BX647643.1 | - | 11 - 12 | 1413 | 69 |