The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032096.1 | NP_115267.1 | 1 - 1 | 2388 | 78 |
GenBank mRNA | AF152518.1 | - | 1 - 1 | 2388 | 78 |
BC101805.1 | - | 1 - 1 | 2451 | 78 |