The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032090.1 | NP_114479.1 | 1 - 1 | 2437 | 157 |
GenBank mRNA | AF152503.1 | - | 1 - 1 | 2437 | 157 |
BC093873.1 | - | 1 - 1 | 2501 | 157 | |
BC093875.1 | - | 1 - 1 | 2501 | 157 |