The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256088.1 | NP_001243017.1 | 1 - 2 | 160 | 69 |
GenBank mRNA | AK314137.1 | - | 1 - 2 | 126 | 69 |