The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001310135.1 | NP_001297064.1 | 31 - 32 | 6204 | 171 |
XM_011537430.2 | XP_011535732.1 | 27 - 28 | 4036 | 171 | |
XM_011537431.2 | XP_011535733.1 | 25 - 26 | 3693 | 171 | |
XM_017021254.1 | XP_016876743.1 | 30 - 31 | 6054 | 171 | |
XM_017021255.1 | XP_016876744.1 | 29 - 30 | 5919 | 171 | |
XM_017021256.1 | XP_016876745.1 | 30 - 31 | 6318 | 171 | |
GenBank mRNA | BC014342.2 | - | 4 - 5 | 611 | 171 |
BC103915.1 | - | 12 - 13 | 1553 | 171 | |
BC103916.2 | - | 12 - 13 | 1553 | 171 | |
BX161415.1 | - | 11 - 12 | 1488 | 171 |