The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 6 - 7 | 622 | 62 |
XM_005271736.4 | XP_005271793.2 | 7 - 8 | 1652 | 62 | |
XM_011543072.2 | XP_011541374.1 | 7 - 8 | 854 | 62 | |
XM_011543073.2 | XP_011541375.2 | 7 - 8 | 590 | 62 | |
XM_017018595.1 | XP_016874084.1 | 7 - 8 | 571 | 62 | |
XM_017018596.1 | XP_016874085.1 | 5 - 6 | 848 | 62 | |
XM_017018597.1 | XP_016874086.1 | 7 - 8 | 1661 | 62 | |
GenBank mRNA | AL833062.2 | - | 5 - 6 | 424 | 62 |
BC051236.1 | - | 1 - 2 | 78 | 62 | |
BC065371.1 | - | 1 - 2 | 78 | 62 | |
EF127492.1 | - | 6 - 7 | 622 | 62 |