The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002358.3 | NP_002349.1 | 2 - 3 | 351 | 144 |
GenBank mRNA | AJ000186.1 | - | 2 - 3 | 293 | 144 |
AK223433.1 | - | 2 - 3 | 312 | 144 | |
AK313827.1 | - | 2 - 3 | 313 | 144 | |
BC000356.2 | - | 2 - 3 | 301 | 144 | |
BC005945.1 | - | 2 - 3 | 294 | 144 | |
BC070283.1 | - | 2 - 3 | 263 | 144 |